Please login to your online account to display your discounted pricing

Sulfuric Acid, 80% (v/v) Aqueous Solution (4 + 1), APHA/EPA Certified Reagent, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
Name Note 80% (v/v) Aqueous Solution (4 + 1)
CAS 7664-93-9
CAS 7732-18-5
Purity Grade Certified APHA/EPA Reagent
Physical Form Liquid
Assay Percent Range 80.0 ± 0.5% (v/v)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R819000020F

 
ricca chemical company
RIR819000020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R81900004A

 
ricca chemical company
RIR81900004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 20L
Purity Grade Certified APHA/EPA Reagent
Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
Solubility Solubility in Water: Infinite
Solvent or Matrix Aqueous Solution
Name Note 80% (v/v) Aqueous Solution (4 + 1)
Assay Percent Range 80.0 ± 0.5% (v/v)
CAS 7732-18-5
Odor Odorless
Packaging Cubitainer