Please login to your online account to display your discounted pricing

Sulfuric Acid, 5N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$104.62 - $552.26

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
Name Note 5N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula H2SO4
Formula Weight 98.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004385

 
avantor performance
569602
Narrow Mouth Polyethylene Bottle 1L Each for $104.62

02004386

 
avantor performance
569603
Cubetainer 4L Each for $552.26
Description & Specifications

Specifications

Formula Weight 98.08
Chemical Name or Material Sulfuric Acid
Molecular Formula H2SO4
Packaging Narrow Mouth Polyethylene Bottle
Name Note 5N Volumetric Solution
Specific Gravity 1L = 1.11kg
Purity Grade BAKER ANALYZED™ Reagent
Quantity 1L