missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Sulfuric Acid, 50% (w/w), Ricca Chemical
Chemical Identifiers
CAS |
7664-93-9 |
Molecular Formula |
H2O4S |
Molecular Weight (g/mol) |
98.07 |
MDL Number |
MFCD00064589 |
InChI Key |
QAOWNCQODCNURD-UHFFFAOYSA-N |
Synonym |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
PubChem CID |
1118 |
ChEBI |
CHEBI:26836 |
IUPAC Name |
sulfuric acid |
SMILES |
OS(O)(=O)=O |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
7664-93-9 |
|
98.07 |
|
QAOWNCQODCNURD-UHFFFAOYSA-N |
|
1118 |
|
sulfuric acid |
|
H2O4S |
|
MFCD00064589 |
|
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
|
CHEBI:26836 |
|
OS(O)(=O)=O |
Specifications
|
7664-93-9,7732-18-5 |
|
500 mL |
|
MFCD00064589 |
|
QAOWNCQODCNURD-UHFFFAOYSA-N |
|
sulfuric acid |
|
1118 |
|
Sulfuric acid |
|
Safety-coated Glass Bottle |
|
H2O4S |
|
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
|
OS(O)(=O)=O |
|
98.07 |
|
CHEBI:26836 |
SDS
missing translation for 'documents'