Please login to your online account to display your discounted pricing

Alfa Aesar™ Sulfuric acid, 5% v/v aq. soln.

Chemical Identifiers

Chemical Name or Material Sulfuric acid
Formula Weight 98.08
MDL Number MFCD00064589
UN Number UN2796
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA35606K2

  SDS
alfa aesar™
35606K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sulfuric acid
Quantity 1L
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H319
Causes severe skin burns and eye damage.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SULFURIC ACID
TSCA TSCA
Physical Form Liquid
Formula Weight 98.08
MDL Number MFCD00064589
UN Number UN2796
Concentration 5% w/v aq. soln.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H319
Causes severe skin burns and eye damage.
Causes serious eye irritation.