missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric Acid, 5.00 Normal, Ricca Chemical

$46.30 - $198.94

Chemical Identifiers

CAS 7732-18-5,7664-93-9
CAS Min % 78.0,19.96
CAS Max % 81.0,20.77
Physical Form Liquid
Chemical Name or Material Sulfuric Acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
8325-1 SDS Ricca Chemical Company
RI83251
Poly natural 4L Each for $198.94
8325-32 SDS Ricca Chemical Company
RI832532
Poly natural 1L Each for $73.58
8325-5PT Ricca Chemical Company
RI83255PT
Glass clr sc 2.50L Each for $120.52
8325-16 SDS Ricca Chemical Company
RI832516
Poly natural 500mL Each for $46.30
8325-5 Ricca Chemical Company
RI83255
Cubitainer 20L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
83255HP Ricca Chemical Company
RI83255HP
Ropak™ 20L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7664-93-9
81.0,20.77
Sulfuric Acid
78.0,19.96
Liquid

Specifications

Clear colorless
7732-18-5,7664-93-9
81.0,20.77
Poly natural
Sulfuric Acid
4L
78.0,19.96
Liquid
5.00Normal