Please login to your online account to display your discounted pricing

Sulfuric Acid, 4.00 Normal, Ricca Chemical

$172.67 - $172.67

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Min % 81.0
CAS Max % 85.0
CAS 7664-93-9
CAS Min % 16.67
CAS Max % 17.35
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

832225

  SDS
ricca chemical company
RI83222.5
Cubitainer 10L Each for $172.67

832216

 
ricca chemical company
RI832216
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

83225PT

 
ricca chemical company
RI83225PT
Glass clr sc 2.50L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sulfuric Acid
Color Clear colorless
Quantity 10L
CAS 7732-18-5
CAS Min % 81.0
CAS Max % 85.0
CAS 7664-93-9
CAS Min % 16.67
CAS Max % 17.35
Physical Form Liquid
Packaging Cubitainer
Concentration or Composition Notes 4.00Normal