Please login to your online account to display your discounted pricing

Sulfuric Acid, 3% (v/v) Aqueous Solution, Ricca Chemical

$689.24 - $689.24

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7664-93-9
CAS Min % 5.13
CAS Max % 5.34
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R81483004A

 
ricca chemical company
RIR81483004A
Poly natural 4L Each for $689.24

R81483001A

  SDS
ricca chemical company
RIR81483001A
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 4L
Physical Form Liquid
Color Clear colorless
Packaging Poly natural
Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
CAS Min % 5.13
CAS Max % 5.34
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
Concentration or Composition (by Analyte or Components) 3% (v/v) Aqueous Solution
Shelf Life 24 months
Safety and Handling

24 months