Please login to your online account to display your discounted pricing

Sulfuric Acid, 3.00 Normal, Ricca Chemical

$66.70 - $170.33

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Min % 85.0
CAS Max % 88.0
CAS 7664-93-9
CAS Min % 13.07
CAS Max % 13.6
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8320-1

  SDS
ricca chemical company
RI83201
Poly natural 4L Each for $170.33

8320-32

  SDS
ricca chemical company
RI832032
Poly natural 1L Each for $66.70

8320-5

 
ricca chemical company
RI83205
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Concentration or Composition Notes 3.00Normal
Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Min % 85.0
CAS Max % 88.0
CAS 7664-93-9
CAS Min % 13.07
CAS Max % 13.6
Packaging Poly natural