Please login to your online account to display your discounted pricing

Sulfuric Acid, 25% (v/v) Aqueous Solution, Ricca Chemical

$173.18 - $650.26

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7664-93-9 (Sulfuric Acid)
CAS 7732-18-5 (Water, Deionized)
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8170-1

  SDS
ricca chemical company
RI81701
Poly natural 4L Each for $173.18

8170-5

  SDS
ricca chemical company
RI81705
Cubitainer 20L Each for $650.26

8170-16

 
ricca chemical company
RI817016
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R81740001A

 
ricca chemical company
RIR81740001A
Natural poly 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R81740004A

 
ricca chemical company
RIR81740004A
Natural poly 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R817400020F

 
ricca chemical company
RIR817400020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R8174000500

 
ricca chemical company
RIR8174000500A
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Solvent or Matrix Aqueous Solution
Concentration or Composition Notes 25% (v/v)
CAS 7664-93-9 (Sulfuric Acid)
Chemical Name or Material Sulfuric Acid
Packaging Poly natural
CAS 7732-18-5 (Water, Deionized)