missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric acid, 20% fuming, 18-24% free SO3, Alfa Aesar™

Manufacturer:  Alfa Aesar 4554336

 View more versions of this product

Catalog No. AA4554336



Specifications

Specifications

Sulfuric acid; 15% fuming
8014-95-7
H2O7S2
oleum, fuming sulfuric acid, sulfuric acid, fuming, oleum iodisum, sulfuric acid fuming, sulfuric acid fuming, oleum fuming sulfuric acid, sulfuric acid, mixt with sulfur trioxide, sulfur trioxide, mixt. with sulfuric acid, sulfuric acid mixture with sulfur trioxide
1.9
sulfuric acid;sulfur trioxide
24681
500g
20% fuming, 18-24% free SO3
MFCD00137792
HIFJUMGIHIZEPX-UHFFFAOYSA-N
OS(=O)(=O)O.O=S(=O)=O
178.129
Liquid

Chemical Identifiers

8014-95-7
178.129
HIFJUMGIHIZEPX-UHFFFAOYSA-N
24681
OS(=O)(=O)O.O=S(=O)=O
H2O7S2
MFCD00137792
oleum, fuming sulfuric acid, sulfuric acid, fuming, oleum iodisum, sulfuric acid fuming, sulfuric acid fuming, oleum fuming sulfuric acid, sulfuric acid, mixt with sulfur trioxide, sulfur trioxide, mixt. with sulfuric acid, sulfuric acid mixture with sulfur trioxide
sulfuric acid;sulfur trioxide
SDS Documents