Please login to your online account to display your discounted pricing

Sulfuric Acid, 2.50N, Ricca Chemical

$47.88 - $147.55

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Min % 87.0
CAS Max % 90.0
CAS 7664-93-9
CAS Min % 11.12
CAS Max % 11.58
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8315-1

  SDS
ricca chemical company
RI83151
Poly natural 4L Each for $147.55

8315-32

  SDS
ricca chemical company
RI831532
Poly natural 1L Each for $47.88

83155

 
ricca chemical company
RI83155
Cubitainer™ 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sulfuric Acid
Quantity 4L
CAS 7732-18-5
CAS Min % 87.0
CAS Max % 90.0
CAS 7664-93-9
CAS Min % 11.12
CAS Max % 11.58
Packaging Poly natural
Concentration or Composition Notes 2.50N