missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric Acid, Certified, 2.00N ±0.02N (1.0M), LabChem™

Supplier:  LabChem LC257902

 View more versions of this product

Catalog No. LC257902



Description

Description

Specifications

Specifications

Sulfuric Acid, 2.0N (1.0M)
7664-93-9,7732-18-5
1.06g/mL
Passes Test
1.06g/mL
Sulfur compounds; Thermal decomposition generates: Corrosive vapors
1.14cSt
Liquid
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.08
Colorless
2.00N ± 0.02N
90.75,9.25
Poly Bottle
Traceable to NIST
1 L
H2O4S
H2SO4
UN2796
Soluble in water
OS(O)(=O)=O
98.07
CHEBI:26836
Certified

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/docto

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions