Please login to your online account to display your discounted pricing

Sulfuric Acid, 1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$78.68 - $545.19

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
Name Note 1N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula H2SO4
Formula Weight 98.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004370

 
avantor performance
564202
Narrow Mouth Polyethylene Bottle 1L Each for $78.68

02004372

 
avantor performance
564203
Cubetainer 4L Each for $545.19
Description & Specifications

Specifications

Formula Weight 98.08
Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
Specific Gravity 1L = 1.04kg
Name Note 1N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula H2SO4
Packaging Narrow Mouth Polyethylene Bottle
Quantity 1L