missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sulfuric Acid, 1.00 N, Ricca Chemical
Supplier: Ricca Chemical Company 830016
Specifications
Sulfuric Acid | |
93.0,4.78 | |
97.0,4.97 | |
500 mL | |
MFCD00064589 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 |
7664-93-9,7732-18-5 | |
1.00N | |
Natural Poly Bottle | |
H2O4S | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 |
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |