missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric acid, 1N (0.5M) standard solution, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 124240010

 View more versions of this product

Catalog No. AC124240010



Description

Description

Volumetric standard solution
Specifications

Specifications

Sulfuric acid
1L
7664-93-9,7732-18-5
H2O4S
MFCD00064589
15, 9104
QAOWNCQODCNURD-UHFFFAOYSA-N
0.998 to 1.002N (20°C)
1.05g/mL
1118
98.07
Pure
Colorless
1N (0.5M) Standard Solution
Vapor Pressure: 1mmHg at 20°C
H2SO4
01,470; 05,633; 06,558; 07,347; 09,441
acide sulfurique, acidum sulfuricum, battery acid, dihydrogen sulfate, dipping acid, electrolyte acid, mattling acid, oil of vitriol, sulphuric acid, vitriol brown oil
OS(=O)(=O)O
sulfuric acid
98.072
CHEBI:26836
Liquid
Plastic bottle

Chemical Identifiers

7664-93-9
98.072
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
acide sulfurique, acidum sulfuricum, battery acid, dihydrogen sulfate, dipping acid, electrolyte acid, mattling acid, oil of vitriol, sulphuric acid, vitriol brown oil
CHEBI:26836
OS(=O)(=O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents
Product Certifications