Please login to your online account to display your discounted pricing

Sulfuric Acid, 16.7% (v/v) Aqueous Solution, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Min % 83.0
CAS Max % 84.0
CAS 7664-93-9
CAS Min % 16.0
CAS Max % 17.0
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8162-16

 
ricca chemical company
RI816216
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

8162-32

 
ricca chemical company
RI816232
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R81620004A

 
ricca chemical company
RIR81620004A
Natural poly 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 500mL
Chemical Name or Material Sulfuric Acid
Concentration or Composition Notes 16.7% (v/v)
Solvent or Matrix Aqueous Solution
CAS 7732-18-5
CAS Min % 83.0
CAS Max % 84.0
CAS 7664-93-9
CAS Min % 16.0
CAS Max % 17.0
Packaging Poly natural