Please login to your online account to display your discounted pricing

Sulfuric Acid, 10.0 Normal, Ricca Chemical

$93.18 - $1,052.55

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Min % 65.0
CAS Max % 67.0
CAS 7664-93-9
CAS Min % 33.21
CAS Max % 34.56
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8340-1

 
ricca chemical company
RI83401
Poly natural 4L Each for $353.55

8340-5

 
ricca chemical company
RI83405
Cubitainer 20L Each for $1,052.55

8340-32

 
ricca chemical company
RI834032
Poly natural 1L Each for $93.18

834016

 
ricca chemical company
RI834016
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

83405PT

 
ricca chemical company
RI83405PT
Glass clr sc 2.50L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R83400004F

 
ricca chemical company
RIR83400004F
Cubitainer 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Concentration or Composition Notes 10.0Normal
Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Min % 65.0
CAS Max % 67.0
CAS 7664-93-9
CAS Min % 33.21
CAS Max % 34.56
Packaging Poly natural