Please login to your online account to display your discounted pricing

Sulfuric Acid, 1 Normal, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
Name Note 1N, TP Acid Digest Reagent
CAS 7664-93-9
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 1.000 ± 0.001 N at 20°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R83000004F

 
ricca chemical company
RIR83000004F
Cubitainer 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R830000055E

 
ricca chemical company
RIR830000055E
Poly Drum 55 gal. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Solubility Solubility in Water: Infinite
CAS 7664-93-9
Name Note 1N, TP Acid Digest Reagent
Solvent or Matrix Aqueous Solution
Packaging Cubitainer
CAS 7732-18-5
Odor Odorless
Chemical Name or Material Sulfuric Acid
Assay Percent Range 1.000 ± 0.001 N at 20°C