missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 7664-93-9 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 98.072 |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
Synonym | acide sulfurique, acidum sulfuricum, battery acid, dihydrogen sulfate, dipping acid, electrolyte acid, mattling acid, oil of vitriol, sulphuric acid, vitriol brown oil |
PubChem CID | 1118 |
ChEBI | CHEBI:26836 |
IUPAC Name | sulfuric acid |
SMILES | OS(=O)(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
83-001-CS
|
Ricca Chemical Company
RIR83000004X4A |
4 x 4L | Poly natural |
Each for $387.91
|
|
83-003-2CS
|
Ricca Chemical Company
RIR830000012X1A |
12 x 1L | Poly natural |
Each for $483.58
|
|
Chemical Identifiers
7664-93-9 | |
98.072 | |
acide sulfurique, acidum sulfuricum, battery acid, dihydrogen sulfate, dipping acid, electrolyte acid, mattling acid, oil of vitriol, sulphuric acid, vitriol brown oil | |
CHEBI:26836 | |
OS(=O)(=O)O |
H2O4S | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
Specifications
Colorless | |
7664-93-9,7732-18-5 | |
H2O4S | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
Liquid | |
Aqueous Solution |
4 x 4L | |
1.000 ± 0.001 N at 20°C | |
acide sulfurique, acidum sulfuricum, battery acid, dihydrogen sulfate, dipping acid, electrolyte acid, mattling acid, oil of vitriol, sulphuric acid, vitriol brown oil | |
OS(=O)(=O)O | |
98.072 | |
CHEBI:26836 | |
Poly natural | |
Sulfuric Acid, 1.00 Normal |