Please login to your online account to display your discounted pricing

Sulfuric Acid, 0.6667 Normal, Suitable for Preparation of Protein-free Filtrates of Blood, Serum, or Plasma, Ricca Chemical

$47.82 - $120.70

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Min % 95.0
CAS Max % 99.0
CAS 7664-93-9
CAS Min % 3.24
CAS Max % 3.37
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8290-1

 
ricca chemical company
RI82901
Poly natural 4L Each for $120.70

8290-32

 
ricca chemical company
RI829032
Poly natural 1L Each for $47.82

8290-5

 
ricca chemical company
RI82905
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sulfuric Acid
Color Clear colorless
Quantity 4L
CAS 7732-18-5
CAS Min % 95.0
CAS Max % 99.0
CAS 7664-93-9
CAS Min % 3.24
CAS Max % 3.37
Physical Form Liquid
Packaging Poly natural
Concentration or Composition Notes 0.6667Normal

Suitable For Preparation of Protein-free Filtrates of Blood, Serum, or Plasma