Please login to your online account to display your discounted pricing

Sulfuric Acid, 0.300 Molar (0.600 Normal), Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
Name Note 0.300 Molar (0.600 Normal), TP Acid Digest Reagent
CAS 7664-93-9
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 0.6000 ± 0.0005 N at 20°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R828500010F

 
ricca chemical company
RIR828500010F
Cubitainer 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R82850001A

 
ricca chemical company
RIR82850001A
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R82850004A

 
ricca chemical company
RIR82850004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 10L
Solvent or Matrix Aqueous Solution
CAS 7664-93-9
CAS 7732-18-5
Packaging Cubitainer
Solubility Solubility in Water: Infinite
Name Note 0.300 Molar (0.600 Normal), TP Acid Digest Reagent
Odor Odorless
Assay Percent Range 0.6000 ± 0.0005 N at 20°C
Chemical Name or Material Sulfuric Acid