Please login to your online account to display your discounted pricing

Sulfuric Acid, 0.2N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$89.75 - $182.59

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
Name Note 0.2N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula H2SO4
Formula Weight 98.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004367

 
avantor performance
569003
Cubetainer 4L Each for $182.59

02004366

 
avantor performance
569002
Narrow Mouth Polyethylene Bottle 1L Each for $89.75
Description & Specifications

Specifications

Formula Weight 98.08
Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
Molecular Formula H2SO4
Purity Grade BAKER ANALYZED™ Reagent
Quantity 4L
Packaging Cubetainer
Specific Gravity 1L = 1.01kg
Name Note 0.2N Volumetric Solution