Please login to your online account to display your discounted pricing

Sulfuric Acid, 0.255N, Ricca Chemical

For Analysis of Suitable For Crude Fiber Analysis of Food and Feed

$93.24 - $220.71

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7664-93-9 (Sulfuric Acid)
CAS 7732-18-5 (Water, Deionized)
Molecular Formula H2SO4
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8272-1

 
ricca chemical company
RI82721
Poly natural 4L Each for $93.24

8272-5

 
ricca chemical company
RI82725
Cubitainer™ 20L Each for $220.71

8272-32

 
ricca chemical company
RI827232
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sulfuric Acid
Quantity 4L
CAS 7664-93-9 (Sulfuric Acid)
CAS 7732-18-5 (Water, Deionized)
Molecular Formula H2SO4
Concentration or Composition (by Analyte or Components) 0.255N (12.5g/L)
Packaging Poly natural