missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sulfuric Acid, 0.250 N (N/4), Ricca Chemical
$82.59 - $425.75
Chemical Identifiers
CAS | 7664-93-9 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 98.07 |
MDL Number | MFCD00064589 |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
PubChem CID | 1118 |
ChEBI | CHEBI:26836 |
IUPAC Name | sulfuric acid |
SMILES | OS(O)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
8270-32
|
Ricca Chemical Company
827032 |
1 L | Natural Poly Bottle |
Each for $82.59
|
|
||||
8270-1
|
Ricca Chemical Company
82701 |
4 L | Natural Poly Bottle |
Each for $196.94
|
|
||||
8270-5
|
Ricca Chemical Company
82705 |
20 L | Cubitainer |
Each for $425.75
|
|
||||
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Specifications
Colorless | |
97.0,1.24 | |
100.0,1.29 | |
1 L | |
Liquid | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 |
7664-93-9,7732-18-5 | |
0.250N | |
Natural Poly Bottle | |
H2O4S | |
MFCD00064589 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
Sulfuric Acid |