Please login to your online account to display your discounted pricing

Sulfuric Acid, 0.250 Normal (N/4), Ricca Chemical


Quantity 4L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR82700004F

 View more versions of this product

Catalog No. R82700004F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 4L
Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
Name Note 0.250 Normal (N/4), TP wash
Solvent or Matrix Aqueous Solution
Odor Odorless
Solubility Solubility in Water: Infinite
Packaging Cubitainer
Assay Percent Range 0.2500 ± 0.0001 N at 20°C
CAS 7664-93-9

Specifications

Chemical Name or Material Sulfuric Acid
Name Note 0.250 Normal (N/4), TP wash
CAS 7664-93-9
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 0.2500 ± 0.0001 N at 20°C