Please login to your online account to display your discounted pricing

Sulfuric Acid, 0.250 Normal (N/4), Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
Name Note 0.250 Normal (N/4), TP wash
CAS 7664-93-9
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 0.2500 ± 0.0001 N at 20°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R82700004C

 
ricca chemical company
RIR82700004C
Amber glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R82700004F

 
ricca chemical company
RIR82700004F
Cubitainer 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R8270000500

 
ricca chemical company
RIR8270000500A
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Chemical Name or Material Sulfuric Acid
Solubility Solubility in Water: Infinite
Solvent or Matrix Aqueous Solution
Assay Percent Range 0.2500 ± 0.0001 N at 20°C
CAS 7664-93-9
CAS 7732-18-5
Packaging Amber glass
Name Note 0.250 Normal (N/4), TP wash
Odor Odorless