Please login to your online account to display your discounted pricing

Sulfuric Acid, 0.1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$85.94 - $559.48

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
Name Note 0.1N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula H2SO4
Formula Weight 98.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004365

 
avantor performance
564103
Cubetainer 4L Each for $559.48

02004363

 
avantor performance
564102
Narrow Mouth Polyethylene Bottle 1L Each for $85.94
Description & Specifications

Specifications

Formula Weight 98.08
Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
Specific Gravity 1L = 1.02kg
Name Note 0.1N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula H2SO4
Packaging Cubetainer
Quantity 4L