Please login to your online account to display your discounted pricing

Sulfuric Acid, 0.125 Normal, Ricca Chemical

$131.64 - $131.64

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 7664-93-9
CAS Min % 0.63
CAS Max % 0.65
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8256-1

 
ricca chemical company
RI82561
Poly natural 4L Each for $131.64

825616

 
ricca chemical company
RI825616
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

825632

 
ricca chemical company
RI825632
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

8256-5

 
ricca chemical company
RI82565
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Concentration or Composition Notes 0.125Normal
CAS 7664-93-9
CAS 7732-18-5
CAS Min % 0.63
CAS Max % 0.65
CAS Min % 97.0
CAS Max % 100.0
Packaging Poly natural
Chemical Name or Material Sulfuric Acid