missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric Acid, 0.1142 N, Ricca Chemical

Suitable for Crude Fiber Analysis of Food and Feed

Supplier:  Ricca Chemical Company 82551

 View more versions of this product

Catalog No. 8255-1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4 L
Sulfuric Acid
97.0,0.57
Natural Poly Bottle
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
Colorless
7664-93-9,7732-18-5
100.0,0.6
H2O4S
OS(=O)(=O)O
98.072
CHEBI:26836

Chemical Identifiers

7664-93-9
98.072
1118
sulfuric acid
H2O4S
QAOWNCQODCNURD-UHFFFAOYSA-N
CHEBI:26836
OS(=O)(=O)O
SDS
Documents

Documents

Promotions

Promotions