Please login to your online account to display your discounted pricing

Sulfuric Acid, 0.100N, Ricca

$34.33 - $404.21

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 7664-93-9
CAS Min % 0.5
CAS Max % 0.52
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8250-1

  SDS
ricca chemical company
RI82501
Poly natural 4L Each for $91.94

8250-32

 
ricca chemical company
RI825032
Poly natural 1L Each for $41.03

825016

 
ricca chemical company
RI825016
Poly natural 500mL Each for $34.33

8250-5

 
ricca chemical company
RI82505
Cubitainer™ 20L Each for $248.58

82-503-2CS

 
ricca chemical company
RIR825000012X1A
Poly natural 12L Each for $404.21

82-501-CS

 
ricca chemical company
RIR82500004X4A
Poly natural 16L Each for $315.00

8250-2-5

 
ricca chemical company
RI82502.5
Cubitainer™ 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 4L
Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
Concentration or Composition (by Analyte or Components) 0.100N
CAS Min % 97.0
CAS Max % 100.0
CAS 7664-93-9
CAS Min % 0.5
CAS Max % 0.52
Packaging Poly natural