Please login to your online account to display your discounted pricing

Sulfuric Acid, 0.0833 Normal, for Preparation of Protein-free Filtrates, Ricca Chemical


Quantity 20L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR823000020F

 View more versions of this product

Catalog No. R823000020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 20L
Chemical Name or Material Sulfuric Acid
Concentration or Composition Notes 0.0833Normal
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 7664-93-9
CAS Min % 0.42
CAS Max % 0.43
Packaging Cubitainer

Specifications

Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 7664-93-9
CAS Min % 0.42
CAS Max % 0.43
Physical Form Liquid