missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric acid, 0.05N Standardized Solution, Alfa Aesar™

Manufacturer:  Alfa Aesar 45048M4

 View more versions of this product

Catalog No. AA45048M4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Sulfuric acid
20L
Colourless solution
MFCD00064589
98.08
Colorless
7732-18-5
H2SO4
dihydrogen oxide, dihydrogen monoxide
Liquid

Chemical Identifiers

7732-18-5
H2SO4
MFCD00064589
dihydrogen oxide, dihydrogen monoxide
SDS Documents