Please login to your online account to display your discounted pricing

Sulfuric Acid, 0.0400 Normal (N/25), 0.0200 Molar (M/50), Ricca Chemical

$40.36 - $93.39

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 7664-93-9
CAS Min % 0.2
CAS Max % 0.21
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8215-32

 
ricca chemical company
RI821532
Poly natural 1L Each for $40.36

8215-1

  SDS
ricca chemical company
RI82151
Poly natural 4L Each for $93.39

R8215000500

 
ricca chemical company
RIR8215000500A
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 1L
Concentration or Composition Notes 0.0400Normal,0.0200Molar
CAS 7732-18-5
Chemical Name or Material Sulfuric Acid
Packaging Poly natural
CAS Min % 98.0
CAS Max % 0.21
CAS Max % 100.0
CAS Min % 0.2
CAS 7664-93-9