Please login to your online account to display your discounted pricing

Sulfuric Acid, 0.0100 Normal, Ricca Chemical


Quantity 4L
Packaging Amber glass

Manufacturer: ricca chemical company  RIR82500004C

 View more versions of this product

Catalog No. R82500004C

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 4L
Concentration or Composition Notes 0.0100Normal
CAS Max % 100.0
Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Min % 98.0
CAS 7664-93-9
CAS Min % 0.05
Packaging Amber glass
CAS Max % 0.05

Specifications

Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 7664-93-9
CAS Min % 0.05
CAS Max % 0.05
Physical Form Liquid