missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sulfur, 99.5+%, refined, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 201220100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For ultra trace analysisSpecifications
Sulfur | |
10kg | |
S | |
01,1118; 03,273; 05,632; 06,556; 14,291; 15,297 | |
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot | |
NINIDFKCEFEMDL-UHFFFAOYSA-N | |
sulfur | |
5362487 | |
32.05 | |
Plastic drum |
7704-34-9 | |
99.5% min | |
168°C | |
15,9099 | |
Solubility in water: insoluble. Other solubilities: slightly soluble in alcohol and ether,soluble in carbon disulfide,benzene,(about 2.4%,at 30°C,much more at higher temps.),toluene,,acetone (2.65% at 25°C),chloroform (1.5% at 18°C) | |
[S] | |
32.05 | |
CHEBI:26833 | |
99.5+% |
Safety and Handling
GHS H Statement
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and w
GHS Signal Word: Warning
EINECSNumber : 231-722-6
RUO â Research Use Only