missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sulfur powder, -325 mesh, 99.5%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 043766A3
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sulphur | |
7704-34-9 | |
2 kg | |
S | |
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot | |
[S] | |
32.06 | |
CHEBI:26833 | |
Pale yellow powder |
Yellow | |
Powder | |
99.5% | |
MFCD00085316 | |
NINIDFKCEFEMDL-UHFFFAOYSA-N | |
sulfur | |
5362487 | |
Weak |
Chemical Identifiers
7704-34-9 | |
32.06 | |
NINIDFKCEFEMDL-UHFFFAOYSA-N | |
5362487 | |
sulfur |
S | |
MFCD00085316 | |
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot | |
CHEBI:26833 | |
[S] |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319-H500
DOTInformation : DOT Class: 4.1, Packaging Group: III
EINECSNumber : 231-722-6
RTECSNumber : WS4250000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only