missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfur pieces, 99.99% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1075530

 View more versions of this product

Catalog No. AA1075530



Specifications

Specifications

Sulphur
99.999% (Metals basis)
MFCD00085316
acmc-20a47y
[S]
32.06
CHEBI:26833
Weak
250g
7704-34-9
S
Pale yellow pieces
NINIDFKCEFEMDL-UHFFFAOYSA-N
sulfur
5362487
Pieces
Yellow

Chemical Identifiers

7704-34-9
32.06
NINIDFKCEFEMDL-UHFFFAOYSA-N
5362487
sulfur
S
MFCD00085316
acmc-20a47y
CHEBI:26833
[S]
Safety and Handling

Safety and Handling

H228-H315: Flammable solid. Causes skin irritation.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-722-6

Documents