missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sulfur in Kerosene standard solution, Specpure™, 500μg/g (0.050%)

$131.04 - $131.04

Chemical Identifiers

MDL Number MFCD00804268
UN Number UN1223
Chemical Name or Material Sulfur in Kerosene standard solution, 500μg/g (0.050%)
Grade Specpure™
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA40539AE SDS Alfa Aesar™
40539AE*
100mL Each for $131.04
Specifications Description & Specifications

Chemical Identifiers

MFCD00804268
UN1223
Sulfur in Kerosene standard solution, 500μg/g (0.050%)
Specpure™

Specifications

MFCD00804268
100mL
Specpure™
UN1223
Sulfur in Kerosene standard solution, 500μg/g (0.050%)

Safety and Handling

GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.

GHS P Statement
P210-P280-P301+P310-P405-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KEROSENE

TSCA : TSCA