Please login to your online account to display your discounted pricing

Alfa Aesar™ Sulfur in Kerosene standard solution, Specpure™, 10μg/g (0.001%)

$134.84 - $134.84

Chemical Identifiers

Chemical Name or Material Sulfur in Kerosene standard solution
Name Note 10μg/g (0.001%)
Purity Grade Specpure™
MDL Number MFCD00804268
UN Number UN1223
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41704AE

  SDS
alfa aesar™
41704AE*
100mL Each for $134.84
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P405-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Sulfur in Kerosene standard solution
Quantity 100mL
TSCA TSCA
Name Note 10μg/g (0.001%)
Purity Grade Specpure™
MDL Number MFCD00804268
UN Number UN1223
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KEROSENE
Safety and Handling

Danger

GHS P Statement
P210-P280-P301+P310-P405-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.