Please login to your online account to display your discounted pricing

Sulfur ICP Standard, 1mL = 1mg S (1,000ppm S)(NH4)2SO4 in H2O, Ricca Chemical

$153.91 - $153.91

Chemical Identifiers

Chemical Name or Material Sulfur ICP Standard
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 7783-20-2
CAS Min % 0.4
CAS Max % 0.42
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PS1KW-100

 
ricca chemical company
RIPS1KW100
Poly natural 100mL Each for $153.91

PS1KW-500

 
ricca chemical company
RIPS1KW500
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 100mL
Concentration or Composition (by Analyte or Components) 1mL = 1mg S (1, 000ppm S)(NH4)2SO4 in H2O
CAS 7732-18-5
CAS Max % 100.0
Shelf Life 18 months
CAS 7783-20-2
CAS Min % 0.4
CAS Min % 98.0
CAS Max % 0.42
Chemical Name or Material Sulfur ICP Standard
Packaging Poly natural
Safety and Handling

18 months