Please login to your online account to display your discounted pricing

Alfa Aesar™ Sulfur in Heavy Mineral Oil standard solution, Specpure™, 50,000μg/g (5.00%)

Chemical Identifiers

Chemical Name or Material Sulfur in Heavy Mineral Oil standard solution
Name Note 50,000μg/g (5.00%)
Purity Grade Specpure™
MDL Number MFCD00804270
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40361AE

  SDS
alfa aesar™
40361AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 2 GHS H Statement
H350-H315-H319
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material Sulfur in Heavy Mineral Oil standard solution
Quantity 100mL
TSCA TSCA
Name Note 50,000μg/g (5.00%)
Purity Grade Specpure™
MDL Number MFCD00804270
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P281-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P280-P281-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H315-H319
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.