Please login to your online account to display your discounted pricing

Alfa Aesar™ Sulfur in Heavy Mineral Oil standard solution, Specpure™, 300μg/g (0.0300%)

Chemical Identifiers

Chemical Name or Material Sulfur in Heavy Mineral Oil standard solution
Name Note 300μg/g (0.0300%)
Purity Grade Specpure™
MDL Number MFCD00804270
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40347AE

 
alfa aesar™
40347AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sulfur in Heavy Mineral Oil standard solution
Quantity 100mL
TSCA TSCA
Name Note 300μg/g (0.0300%)
Purity Grade Specpure™
MDL Number MFCD00804270
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H350
May cause cancer.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350
May cause cancer.