Please login to your online account to display your discounted pricing

Alfa Aesar™ Sulfur in Diesel Fuel standard solution, Specpure™, 300μg/g (0.030%)

Chemical Identifiers

Chemical Name or Material Sulfur in Diesel Fuel standard solution
Name Note 300μg/g (0.030%)
Purity Grade Specpure™
MDL Number MFCD01095233
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38767AE

 
alfa aesar™
38767AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sulfur in Diesel Fuel standard solution
Quantity 100mL
TSCA TSCA
Name Note 300μg/g (0.030%)
Purity Grade Specpure™
MDL Number MFCD01095233
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351-H315-H319-H335-H227
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H315-H319-H335-H227
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.