Learn More
Sulfur in Diesel Fuel standard solution, Specpure™, 200μg/g (0.020%), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 038766AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sulfur in Diesel Fuel standard solution | |
100 mL | |
Specpure™ |
200μg/g (0.020%) | |
MFCD01095233 |
Chemical Identifiers
MFCD01095233 |
Safety and Handling
GHS H Statement
H351-H315-H319-H335-H227
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P201-P202-P210-P235-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335-H351
EINECSNumber : 270-676-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only