Please login to your online account to display your discounted pricing

Alfa Aesar™ Sulfur in Diesel Fuel standard solution, Specpure™, 20,000μg/g (2.00%)

$140.74 - $140.74

Chemical Identifiers

Chemical Name or Material Sulfur in Diesel Fuel standard solution
Name Note 20,000μg/g (2.00%)
Purity Grade Specpure™
MDL Number MFCD01095233
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38778AE

  SDS
alfa aesar™
38778AE
100mL Each for $140.74
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD01095233
Quantity 100mL
Name Note 20,000μg/g (2.00%)
Purity Grade Specpure™
Health Hazard 1 Warning
TSCA TSCA
Health Hazard 2 GHS H Statement
H351-H315-H319-H335-H227
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Chemical Name or Material Sulfur in Diesel Fuel standard solution
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H315-H319-H335-H227
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.