Please login to your online account to display your discounted pricing

Alfa Aesar™ Sulfur in Diesel Fuel standard solution, Specpure™, 1000μg/g (0.10%)

$137.42 - $137.42

Chemical Identifiers

Chemical Name or Material Sulfur in Diesel Fuel standard solution
Name Note 1000μg/g (0.10%)
Purity Grade Specpure™
MDL Number MFCD01095233
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38771AE

  SDS
alfa aesar™
38771AE
100mL Each for $137.42
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P210-P280-P281-P308+P313-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Name Note 1000μg/g (0.10%)
TSCA TSCA
Chemical Name or Material Sulfur in Diesel Fuel standard solution
Health Hazard 2 GHS H Statement
H351-H227
Suspected of causing cancer.
Combustible liquid.
Quantity 100mL
MDL Number MFCD01095233
Health Hazard 1 Warning
Color Yellow
Odor Petroleum-like
Purity Grade Specpure™
Physical Form Liquid
Safety and Handling

Warning

GHS P Statement
P210-P280-P281-P308+P313-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H227
Suspected of causing cancer.
Combustible liquid.