missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sulfur in Diesel Fuel standard solution, Specpure™, 10,000μg/g (1.00%)

$143.78 - $143.78

Chemical Identifiers

MDL Number MFCD01095233
Chemical Name or Material Sulfur in Diesel Fuel standard solution, 10,000μg/g (1.00%)
Grade Specpure™
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA38776AE SDS Alfa Aesar™
38776AE
100mL Each for $143.78
Specifications Description & Specifications

Chemical Identifiers

MFCD01095233
Sulfur in Diesel Fuel standard solution, 10,000μg/g (1.00%)
Specpure™

Specifications

MFCD01095233
100mL
Sulfur in Diesel Fuel standard solution, 10,000μg/g (1.00%)
Specpure™

Safety and Handling

GHS H Statement
H351-H315-H319-H335-H227
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA