missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfosalicylic Acid Dihydrate (Fine White Crystals), Fisher BioReagents

Manufacturer:  Fisher BioReagents FLBP177500

Catalog No. BP177-500




Description

Description

Sulfosalicylic Acid is used for fixing proteins in agarose and polyacrylamide gels.
Specifications

Specifications

Sulfosalicylic Acid Dihydrate
0.5
106°C
Pass Test
C9H14O6S
MFCD00007508,MFCD00149540
15, 9094
500g
NFYHZVWMQHQKRU-UHFFFAOYSA-N
2-hydroxy-5-sulfobenzoic acid;dihydrate
2723734
Powder/Solid
5965-83-3
White
0.1% max.
99.0 to 101.0 %
HOC6H3(COOH)SO3H·2H2O
Glass Bottle
2-hydroxy-5-sulfobenzoic acid dihydrate, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylic acid dihydrate, 5-sulfosalicylsaeure, 5-sulfosalicylsyre, acmc-1aymd, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, salicylic acid, 5-sulfo-, dihydrate, sulfosalicylic acid dihydrate, unii-09ngq462s6
0.02% max. Insoluble Matter
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
250.27
254.22
99.0 to 101.0%

Chemical Identifiers

5965-83-3
250.27
NFYHZVWMQHQKRU-UHFFFAOYSA-N
2723734
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
C9H14O6S
MFCD00007508,MFCD00149540
2-hydroxy-5-sulfobenzoic acid dihydrate, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylic acid dihydrate, 5-sulfosalicylsaeure, 5-sulfosalicylsyre, acmc-1aymd, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, salicylic acid, 5-sulfo-, dihydrate, sulfosalicylic acid dihydrate, unii-09ngq462s6
2-hydroxy-5-sulfobenzoic acid;dihydrate
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS
Documents
Product Certifications
Promotions

Promotions