Please login to your online account to display your discounted pricing

Sulfosalicylic Acid, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$556.17 - $556.17

Chemical Identifiers

Chemical Name or Material Sulfosalicylic Acid
CAS 5965-83-3
Name Note Dihydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula 2-HOC6H3-1-COOH-5-SO3H·2H2O
Formula Weight 254.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004437

 
avantor performance
036401
Wide Mouth Amber Glass Bottle 500g Each for $556.17
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 254.22
Physical Form Crystals
Chemical Name or Material Sulfosalicylic Acid
CAS 5965-83-3
Molecular Formula 2-HOC6H3-1-COOH-5-SO3H·2H2O
Packaging Wide Mouth Amber Glass Bottle
Name Note Dihydrate
Quantity 500g