Please login to your online account to display your discounted pricing

Sulfosalicylic Acid, Dihydrate, ACS Grade, 99.0 to 101.0%, LabChem™

$213.91 - $213.91

Chemical Identifiers

Chemical Name or Material Sulfosalicylic Acid
CAS 5965-83-3
CAS Max % 100
Name Note Dihydrate
Purity Grade ACS Grade
Linear Formula C7H6O6S·2H2O
Formula Weight 254.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC255201

 
lab chem
LC255201
Amber Glass 500g Each for $213.91
Description & Specifications

Specifications

Formula Weight 254.21
Linear Formula C7H6O6S·2H2O
Solubility Information Soluble in water
Assay 99.0 to 101.0%
Health Hazard 1 Warning
Purity Grade ACS Grade
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Amber Glass
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Name Note Dihydrate
CAS 5965-83-3
CAS Max % 100
Quantity 500g
Recommended Storage Room Temperature
Melting Point 108°C
Physical Form Crystals
Color White
Identification Passes Test
Chemical Name or Material Sulfosalicylic Acid
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Room Temperature